MilliporeSigma
  • Arp2/3-dependent mechanical control of morphogenetic robustness in an inherently challenging environment.
Arp2/3-dependent mechanical control of morphogenetic robustness in an inherently challenging environment.
Developmental cell (2021-02-04)
Emmanuel Martin, Sophie Theis, Guillaume Gay, Bruno Monier, Christian Rouvière, Magali Suzanne
ABSTRACT
Epithelial sheets undergo highly reproducible remodeling to shape organs. This stereotyped morphogenesis depends on a well-defined sequence of events leading to the regionalized expression of developmental patterning genes that finally triggers downstream mechanical forces to drive tissue remodeling at a pre-defined position. However, how tissue mechanics controls morphogenetic robustness when challenged by intrinsic perturbations in close proximity has never been addressed. Using Drosophila developing leg, we show that a bias in force propagation ensures stereotyped morphogenesis despite the presence of mechanical noise in the environment. We found that knockdown of the Arp2/3 complex member Arpc5 specifically affects fold directionality while altering neither the developmental nor the force generation patterns. By combining in silico modeling, biophysical tools, and ad hoc genetic tools, our data reveal that junctional myosin II planar polarity favors long-range force channeling and ensures folding robustness, avoiding force scattering and thus isolating the fold domain from surrounding mechanical perturbations.
Sigma-Aldrich
Schneider′s Insect Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for insect cell culture
Sigma-Aldrich
20-Hydroxyecdysone, ≥93% (HPLC), powder