MilliporeSigma
Water-soluble constituents of ajowan.
Chemical & pharmaceutical bulletin (2001-07-18)
T Ishikawa, Y Sega, J Kitajima
ABSTRACT
From the water-soluble portion of the methanol extract of the fruit of Carum ajowan (ajowan), which has been used as a spice and medicine, 25 compounds, including five new monoterpenoid glucosides, a new monoterpenoid, two new aromatic compound glucosides, and two new glucides, were obtained. Their structures were clarified by spectral investigation.
Sigma-Aldrich
2-C-Methyl-D-erythritol, ≥90% (GC)