MilliporeSigma
Role of Stat3 in lipopolysaccharide-induced IL-10 gene expression.
Journal of immunology (Baltimore, Md. : 1950) (2000-07-21)
E M Benkhart, M Siedlar, A Wedel, T Werner, H W Ziegler-Heitbrock
ABSTRACT
IL-10 is a unique cytokine because it is anti-inflammatory and immunosuppressive. IL-10 is regulated at the level of transcription, but the critical motifs and the relevant transcription factors controlling this gene have remained elusive to date. We now report that a sequence at -120 bp in the human IL-10 promoter binds Stat3 but no other Stat proteins. Mutation of this motif abrogates LPS-induced trans-activation. Overexpression of dominant negative Stat3 suppresses promoter activity, while wild-type Stat3 leads to an enhancement of this activity. Our results show that Stat3, by binding to a single motif in the IL-10 promoter, is controlling expression of the human IL-10 gene.
Sigma-Aldrich
Leupeptin trifluoroacetate salt, ≥90% (HPLC), microbial
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Calpain Inhibitor I, ≥97% (TLC), powder
Sigma-Aldrich
Aprotinin from bovine lung, saline solution, 3-7 TIU/mg protein
Sigma-Aldrich
Pepstatin A, microbial, ≥75% (HPLC)