MilliporeSigma
  • Coordinated expression of beta-amyloid precursor protein and the putative beta-secretase BACE and alpha-secretase ADAM10 in mouse and human brain.
Coordinated expression of beta-amyloid precursor protein and the putative beta-secretase BACE and alpha-secretase ADAM10 in mouse and human brain.
Journal of neurochemistry (2000-10-18)
M Marcinkiewicz, N G Seidah
ABSTRACT
To define the enzymes involved in the etiology of Alzheimer's disease, we compared in mouse and human brain the mRNA levels and cellular localization of the ubiquitous beta-amyloid precursor protein (beta-APP) with those of the putative alpha-secretases ADAM10 and ADAM17 and the beta-secretases BACE and BACE2. In situ hybridization performed in mice during prenatal and postnatal development and in adulthood revealed the coexpression of beta-APP, BACE, and ADAM10. The patterns of BACE2 and ADAM17 only partially overlapped with that of beta-APP. beta-APP, BACE, and ADAM10 mRNAs have also been detected by northern blot in human brain cortex of normal subjects and in Alzheimer's disease subjects. In situ hybridization performed using combined biotin- and radiolabeled riboprobes provided evidence for the coexpression of beta-APP with BACE and ADAM10 in human cortical neurons. Our data provide cytochemical evidence supporting the role of ADAM10 and BACE as authentic alpha- and beta-secretases.