MilliporeSigma
  • Quinacrine Ameliorates Cisplatin-Induced Renal Toxicity via Modulation of Sirtuin-1 Pathway.
Quinacrine Ameliorates Cisplatin-Induced Renal Toxicity via Modulation of Sirtuin-1 Pathway.
International journal of molecular sciences (2021-10-14)
Nada F Abo El-Magd, Hasnaa Ali Ebrahim, Mohamed El-Sherbiny, Nada H Eisa
ABSTRACT
Renal toxicity is a serious side effect that hinders the use of cisplatin, a commonly used and effective chemotherapeutic agent. Meanwhile, quinacrine is an FDA approved drug that has been stated for its anti-inflammatory effect. Thus, we investigated the ameliorative effect of quinacrine against cisplatin-induced renal toxicity. Single intraperitoneal (i.p.) 10 mg/kg cisplatin administration induced renal injury in rats. Our results showed that 10 mg/kg/day quinacrine decreased the mortality rate of rats from 46.15% (cisplatin group) to 12.5%, and significantly decreased renal tissue fibrosis, relative kidney to body weight ratio, serum creatinine and urea levels compared with the cisplatin group. Indeed, quinacrine significantly decreased renal malondialdehyde concentration and increased renal total antioxidant capacity, compared with the cisplatin group. Furthermore, quinacrine caused significant upregulation of renal sirtuin-1 (SIRT-1) with significant downregulation of intercellular adhesion molecule-1 (ICAM-1) and tumor necrosis factor-α (TNF-α). Moreover, quinacrine significantly blocked cisplatin-induced apoptosis, which was made evident by downregulating renal apoptotic proteins (BAX and p53) and upregulating the renal anti-apoptotic protein BCL2, compared with the cisplatin group. In conclusion, this study demonstrates, for the first time, that quinacrine alleviates cisplatin-induced renal toxicity via upregulating SIRT-1, downregulating inflammatory markers (ICAM-1 and TNF-α), reducing oxidative stress, and inhibiting apoptosis.
Sigma-Aldrich
Cisplatin, Cisplatin - CAS 15663-27-1, is a platinum coordination complex with potent anti-neoplastic activity. Induces apoptosis in cancer cells, possibly via caspase-3 activation.
Sigma-Aldrich
Quinacrine dihydrochloride, ≥90%