MilliporeSigma
  • Immunizations with diverse sarbecovirus receptor-binding domains elicit SARS-CoV-2 neutralizing antibodies against a conserved site of vulnerability.
Immunizations with diverse sarbecovirus receptor-binding domains elicit SARS-CoV-2 neutralizing antibodies against a conserved site of vulnerability.
Immunity (2021-11-18)
Deborah L Burnett, Katherine J L Jackson, David B Langley, Anupria Aggrawal, Alberto Ospina Stella, Matt D Johansen, Harikrishnan Balachandran, Helen Lenthall, Romain Rouet, Gregory Walker, Bernadette M Saunders, Mandeep Singh, Hui Li, Jake Y Henry, Jennifer Jackson, Alastair G Stewart, Franka Witthauer, Matthew A Spence, Nicole G Hansbro, Colin Jackson, Peter Schofield, Claire Milthorpe, Marianne Martinello, Sebastian R Schulz, Edith Roth, Anthony Kelleher, Sean Emery, Warwick J Britton, William D Rawlinson, Rudolfo Karl, Simon Schäfer, Thomas H Winkler, Robert Brink, Rowena A Bull, Philip M Hansbro, Hans-Martin Jäck, Stuart Turville, Daniel Christ, Christopher C Goodnow
ABSTRACT
Viral mutations are an emerging concern in reducing SARS-CoV-2 vaccination efficacy. Second-generation vaccines will need to elicit neutralizing antibodies against sites that are evolutionarily conserved across the sarbecovirus subgenus. Here, we immunized mice containing a human antibody repertoire with diverse sarbecovirus receptor-binding domains (RBDs) to identify antibodies targeting conserved sites of vulnerability. Antibodies with broad reactivity against diverse clade B RBDs targeting the conserved class 4 epitope, with recurring IGHV/IGKV pairs, were readily elicited but were non-neutralizing. However, rare class 4 antibodies binding this conserved RBD supersite showed potent neutralization of SARS-CoV-2 and all variants of concern. Structural analysis revealed that the neutralizing ability of cross-reactive antibodies was reserved only for those with an elongated CDRH3 that extends the antiparallel beta-sheet RBD core and orients the antibody light chain to obstruct ACE2-RBD interactions. These results identify a structurally defined pathway for vaccine strategies eliciting escape-resistant SARS-CoV-2 neutralizing antibodies.
Sigma-Aldrich
SIGMAFAST p-Nitrophenyl phosphate Tablets, tablet, To prepare 5 mL
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, commercial grade, powder