MilliporeSigma
  • A nuclease- and bisulfite-based strategy captures strand-specific R-loops genome-wide.
A nuclease- and bisulfite-based strategy captures strand-specific R-loops genome-wide.
eLife (2021-02-24)
Phillip Wulfridge, Kavitha Sarma
ABSTRACT
R-loops are three-stranded nucleic acid structures with essential roles in many nuclear processes. However, their unchecked accumulation is associated with genome instability and is observed in neurodevelopmental diseases and cancers. Genome-wide profiling of R-loops in normal and diseased cells can help identify locations of pathogenic R-loops and advance efforts to attenuate them. We present an antibody-independent R-loop detection strategy, BisMapR, that combines nuclease-based R-loop isolation with non-denaturing bisulfite chemistry to produce genome-wide profiles that retain strand information. BisMapR achieves greater resolution and is faster than existing strand-specific R-loop profiling strategies. In mouse embryonic stem cells, we apply BisMapR to find that gene promoters form R-loops in both directions and uncover a subset of active enhancers that, despite being bidirectionally transcribed, form R-loops exclusively on one strand. BisMapR reveals a previously unnoticed feature of active enhancers and provides a tool to systematically examine their mechanisms in gene expression.
Sigma-Aldrich
GSK-3 Inhibitor XVI, GSK-3 Inhibitor XVI - CAS 252917-06-9, is a cell-permeable, potent, ATP-competitive, and highly selective GSK-3 inhibitor (IC50 = 10 and 6.7 nM against GSK-3α and GSK-3β, respectively).
Sigma-Aldrich
MEK1/2 Inhibitor III, The MEK1/2 Inhibitor III, also referenced under CAS 391210-10-9, controls the biological activity of MEK1/2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.