MilliporeSigma
  • Keratin 19 interacts with GSK3β to regulate its nuclear accumulation and degradation of cyclin D3.
Keratin 19 interacts with GSK3β to regulate its nuclear accumulation and degradation of cyclin D3.
Molecular biology of the cell (2021-08-19)
Pooja Sharma, Sarah Tiufekchiev, Victoria Lising, Seung Woo Chung, Jung Soo Suk, Byung Min Chung
ABSTRACT
Cyclin D3 regulates the G1/S transition and is frequently overexpressed in several cancer types including breast cancer, where it promotes tumor progression. Here we show that a cytoskeletal protein keratin 19 (K19) physically interacts with a serine/threonine kinase GSK3β and prevents GSK3β-dependent degradation of cyclin D3. The absence of K19 allowed active GSK3β to accumulate in the nucleus and degrade cyclin D3. Specifically, the head (H) domain of K19 was required to sustain inhibitory phosphorylation of GSK3β Ser9, prevent nuclear accumulation of GSK3β, and maintain cyclin D3 levels and cell proliferation. K19 was found to interact with GSK3β and K19-GSK3β interaction was mapped out to require Ser10 and Ser35 residues on the H domain of K19. Unlike wildtype K19, S10A and S35A mutants failed to maintain total and nuclear cyclin D3 levels and induce cell proliferation. Finally, we show that the K19-GSK3β-cyclin D3 pathway affected sensitivity of cells toward inhibitors to cyclin-dependent kinase 4 and 6 (CDK4/6). Overall, these findings establish a role for K19 in the regulation of GSK3β-cyclin D3 pathway and demonstrate a potential strategy for overcoming resistance to CDK4/6 inhibitors.
Sigma-Aldrich
Okadaic Acid, Ammonium Salt, Water-soluble analog of Okadaic Acid. Inhibits protein phosphatases 1 and 2A.