MilliporeSigma
  • Genome-Wide Analysis, Modeling, and Identification of Amino Acid Binding Motifs Suggest the Involvement of GH3 Genes during Somatic Embryogenesis of Coffea canephora.
Genome-Wide Analysis, Modeling, and Identification of Amino Acid Binding Motifs Suggest the Involvement of GH3 Genes during Somatic Embryogenesis of Coffea canephora.
Plants (Basel, Switzerland) (2021-10-24)
Hugo A Méndez-Hernández, Ana O Quintana-Escobar, Miguel A Uc-Chuc, Víctor M Loyola-Vargas
ABSTRACT
Auxin plays a central role in growth and plant development. To maintain auxin homeostasis, biological processes such as biosynthesis, transport, degradation, and reversible conjugation are essential. The Gretchen Hagen 3 (GH3) family genes codify for the enzymes that esterify indole-3-acetic acid (IAA) to various amino acids, which is a key process in the induction of somatic embryogenesis (SE). The GH3 family is one of the principal families of early response to auxin genes, exhibiting IAA-amido synthetase activity to maintain optimal levels of free auxin in the cell. In this study, we carried out a systematic identification of the GH3 gene family in the genome of Coffea canephora, determining a total of 18 CcGH3 genes. Analysis of the genetic structures and phylogenetic relationships of CcGH3 genes with GH3 genes from other plant species revealed that they could be clustered in two major categories with groups 1 and 2 of the GH3 family of Arabidopsis. We analyzed the transcriptome expression profiles of the 18 CcGH3 genes using RNA-Seq analysis-based data and qRT-PCR during the different points of somatic embryogenesis induction. Furthermore, the endogenous quantification of free and conjugated indole-3-acetic acid (IAA) suggests that the various members of the CcGH3 genes play a crucial role during the embryogenic process of C. canephora. Three-dimensional modeling of the selected CcGH3 proteins showed that they consist of two domains: an extensive N-terminal domain and a smaller C-terminal domain. All proteins analyzed in the present study shared a unique conserved structural topology. Additionally, we identified conserved regions that could function to bind nucleotides and specific amino acids for the conjugation of IAA during SE in C. canephora. These results provide a better understanding of the C. canephora GH3 gene family for further exploration and possible genetic manipulation.
Sigma-Aldrich
myo-Inositol, ≥99%
Sigma-Aldrich
Kinetin, suitable for plant cell culture, crystalline