MilliporeSigma
  • Parp1-Dependent DNA Double-Strand Break Repair in Irradiated Late Pachytene Spermatocytes.
Parp1-Dependent DNA Double-Strand Break Repair in Irradiated Late Pachytene Spermatocytes.
DNA and cell biology (2020-12-19)
Emad A Ahmed, Abdullah M Alzahrani, Harry Scherthan
ABSTRACT
Poly (ADP-ribose) polymerase-1 (Parp1) is a member of nuclear enzymes family involved in to the response to genotoxic stresses, DNA repair, and is critical for the maintenance of genome stability. During gametogenesis, genome stability is essential for inheritance and formation of healthy gametes. The latter involves DNA double-strand break (DSB)-driven pairing of homologous chromosomes in first meiotic prophase. By analysis of DSB repair kinetics in male meiotic prophase cells of homologous recombination (HR) and nonhomologous end joining (NHEJ)-deficient mouse models, we previously demonstrated an interplay between HR and the conventional NHEJ repair pathway. In the current work, we evaluate the relative contribution of Parp1-dependent NHEJ to the repair of ectopic ionizing radiation (IR)-induced DSBs in control and Parp1-inhibited mouse pachytene spermatocytes before and after the completion of meiotic recombination in stages VI-XI. The disappearance of large, exogenous DSB-related γ-H2AX foci was quantified 1 and 8 h after 1 Gy γ-irradiation of control and 3,4-dihydro-5-[4-(1-piperidinyl)butoxy]-1(2H)quinolinone (DPQ) Parp1-inhibited mice. Late pachytene control spermatocytes obtained 8 h after IR had repaired >80% of DSBs observed at 1 h after IR. However, only 64% of DSBs were repaired in late spermatocytes of DPQ-treated (Parp1-inhibited) mice. Thus, it appears that Parp1 contributes to the repair of a fraction of DSBs in late prophase I, providing further insights in DNA repair pathway choreography during spermatogenic differentiation.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, FITC conjugate, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
DPQ, ≥98% (HPLC), solid