MilliporeSigma
NY-ESO-1 expression predicts an aggressive phenotype of ovarian cancer.
Gynecologic oncology (2017-04-11)
J Brian Szender, Antonios Papanicolau-Sengos, Kevin H Eng, Anthony J Miliotto, Amit A Lugade, Sacha Gnjatic, Junko Matsuzaki, Carl D Morrison, Kunle Odunsi
ABSTRACT
NY-ESO-1 is a cancer testis antigen and a promising target for immunotherapy. The purpose of this study was to determine the expression frequency, immunogenicity, and clinical impact of NY-ESO-1 in ovarian cancer. Immunohistochemistry (IHC), reverse-transcription polymerase chain reaction (RT-PCR), and quantitative-PCR (qRT-PCR) were utilized in an ovarian cancer (including Fallopian tube and primary peritoneal cancers) patient cohort; humoral responses against NY-ESO-1 were determined by ELISA. Clinicopathologic outcomes including progression-free (PFS) and overall (OS) survival were evaluated based on NY-ESO-1 expression. Cohen's kappa (κ) tested agreement between expression tests. NY-ESO-1 expression was detected by any method in 40.7% of 1002 patients' tumors (NY-ESO-1+) and baseline humoral response was identified in 19.0% of 689 tested patients. NY-ESO-1+ patients were older (p<0.001), higher stage (85% stage III/IV vs. 76.4%, p=0.015), less likely to have a complete response to initial therapy (53.9% vs. 68.9%, p=0.002), had more serous histotype (74.5% vs. 66.9%, p=0.011), and had more grade 3 tumors (83.7% vs. 70.8%, p<0.001). There was a trend towards shorter PFS (22.2 vs. 25.0months, p=0.07) and significantly shorter OS (42.9 vs. 50.0months, p=0.003) among NY-ESO-1+ patients. A subset analysis of NY-ESO-1+ patients that received immunotherapy demonstrated improved OS by >2years (52.6 vs. 27.2months, p<0.001). This study is the first demonstration of an association between NY-ESO-1 expression and an aggressive cancer phenotype. The relatively high expression frequency of NY-ESO-1 in ovarian cancer patients coupled with the poor clinical outcomes in NY-ESO-1+ patients reveals an underappreciated need for targeted therapy against this antigen. In support, our study reveals that NY-ESO-1+ patients enrolled on immunotherapy trials targeting the antigen exhibited an improvement in OS.
Sigma-Aldrich
Esculin hydrate, ≥98.0% (HPLC)