MilliporeSigma
  • Distribution of tau hyperphosphorylation in canine dementia resembles early Alzheimer's disease and other tauopathies.
Distribution of tau hyperphosphorylation in canine dementia resembles early Alzheimer's disease and other tauopathies.
Brain pathology (Zurich, Switzerland) (2020-08-19)
Ajantha Abey, Danielle Davies, Claire Goldsbury, Michael Buckland, Michael Valenzuela, Thomas Duncan
ABSTRACT
Some aged community dogs acquire a degenerative syndrome termed Canine Cognitive Dysfunction (CCD) that resembles human dementia because of Alzheimer's Disease (AD), with comparable cognitive and behavioral deficits. Dogs also have similar neuroanatomy, share our domestic environment and develop amyloid-β plaques, making them likely a valuable ecological model of AD. However, prior investigations have demonstrated a lack of neurofibrillary tau pathology in aged dogs, an important hallmark of AD, though elevated phosphorylated tau (p-tau) at the Serine 396 (S396) epitope has been reported in CCD. Here using enhanced immunohistochemical methods, we investigated p-tau in six CCD brains and six controls using the AT8 antibody (later stage neurofibrillary pathology), and an antibody against S396 p-tau (earlier stage tau dysfunction). For the first time, we systematically assessed the Papez circuit and regions associated with Braak staging and found that all CCD dogs displayed elevated S396 p-tau labeling throughout the circuit. The limbic thalamus was particularly implicated, with a similar labeling pattern to that reported for AD neurofibrillary pathology, especially the anterior nuclei, while the hippocampus exhibited dysfunction confined to synaptic layers and efferent pathways. The cingulate and temporal lobes displayed significantly greater tauopathy than the frontal and occipital cortices, also reflective of early Braak staging patterns in AD. Immunofluorescence confirmed that S396 was accumulating within neuronal axons, somata and oligodendrocytes. We also observed AT8 labeling in one CCD brain, near the transentorhinal cortex in layer II neurons, one of the first regions to be affected in AD. Together, these data demonstrate a concordance in regional distribution of tauopathy between CCD and AD, most evident in the limbic thalamus, an important step in further validating CCD as a translational model for human AD and understanding early AD pathogenic mechanisms.
Sigma-Aldrich
Anti-O4 Antibody, clone 81, Alexa Fluor488 Conjugate, clone 81 (mAB 04), from mouse, ALEXA FLUOR 488
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture