MilliporeSigma
  • Neuregulin-1, in a Conducive Milieu with Wnt/BMP/Retinoic Acid, Prolongs the Epicardial-Mediated Cardiac Regeneration Capacity of Neonatal Heart Explants.
Neuregulin-1, in a Conducive Milieu with Wnt/BMP/Retinoic Acid, Prolongs the Epicardial-Mediated Cardiac Regeneration Capacity of Neonatal Heart Explants.
Journal of stem cells & regenerative medicine (2021-08-27)
Himanshu Arora, Alessia C Lavin, Wayne Balkan, Joshua M Hare, Ian A White
ABSTRACT
Rationale: Cardiac sympathetic nerves are required for endogenous repair of the mammalian neonatal heart in vivo, but the underlying mechanism is unclear. Objective: We tested the hypothesis that a combination of cardiac developmental growth factors Wnt3a, BMP4 and Neuregulin (NRG-1), compensate for denervation and support cardiac regeneration in explanted neonatal mammalian hearts. Methods and Results: Hearts from 2-day old neonatal mice were harvested, lesioned at the apex and grown ex vivo for 21 days under defined conditions. Hearts grown in canonical cardiomyocyte culture media underwent complete coagulative necrosis, a process resembling ischemic cell death, by day 14. However, the addition of Wnt3a, BMP-4 and NRG-1, maintained cellular integrity and restored the endogenous regenerative program. None of these factors alone, or in any paired combination, were sufficient to induce regeneration in culture. rNRG-1 alone significantly reduced the accumulation of double strand DNA damage at Day 3; (-NRG-1: 60±12%; +NRG-1: 8±3%; P<0.01) and prevented coagulative necrosis at Day 14. Short-term addition of rWnt3a and rBMP-4 (day 0-3, NRG-1+) increased WT1 expression (a marker of epicardial cells) 7-fold, epicardial proliferation (78±17 cells vs. 21±9 cells; P<0.05), migration and recellularization (80±22 vs. zero cells; P<0.01; n=6) at the injury site on day 14. Conclusions: A novel explant culture system maintains three-dimensional neonatal mouse hearts and the mammalian neonatal cardiac regenerative program ex vivo. We identified that rNRG-1, plus short-term activation of Wnt- and BMP-signaling, promotes cardiac repair via epicardial cell activation, their proliferation and migration to the injury site, followed by putative cardiomyocyte recruitment. This novel technique will facilitate future studies of mammalian cardiac regeneration and may be useful in cardiac-specific drug testing.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-ISL-1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-BrdU Antibody, clone PRB-1, FITC conjugated, clone PRB-1, Chemicon®, from mouse