MilliporeSigma
  • Mycobacterium tuberculosis antigen 85B and ESAT-6 expressed as a recombinant fusion protein in Mycobacterium smegmatis elicits cell-mediated immune response in a murine vaccination model.
Mycobacterium tuberculosis antigen 85B and ESAT-6 expressed as a recombinant fusion protein in Mycobacterium smegmatis elicits cell-mediated immune response in a murine vaccination model.
Molecular immunology (2013-01-22)
Anthony G Tsolaki, Judit Nagy, Sergio Leiva, Uday Kishore, Ida Rosenkrands, Brian D Robertson
ABSTRACT
In this study, we investigated the potential molecular and immunological differences of a recombinant fusion protein (Hybrid-1), comprising of the immunodominant antigens Ag85B and ESAT-6 from Mycobacterium tuberculosis, derived from two different expression systems, namely Mycobacterium smegmatis and Escherichia coli. The fusion protein was successfully expressed and purified from both bacterial hosts and analyzed for any host-dependent post-translational modifications that might affect the immunogenicity of the protein. We investigated the immunogenicity of Hybrid-1 expressed in the two host species in a murine vaccination model, together with a reference standard Hybrid-1 (expressed in E. coli) from the Statens Serum Institut. No evidence of any post-translation modification was found in the M. smegmatis-derived Hybrid-1 fusion protein, nor were there any significant differences in the T-cell responses obtained to the three antigens analyzed. In conclusion, the Hybrid-1 fusion protein was successfully expressed in a homologous expression system using M. smegmatis and this system is worth considering as a primary source for vaccination trials, as it provided protein of excellent yield, stability and free from lipopolysaccharide.
Sigma-Aldrich
Glycoprotein Detection Kit, Sufficient for 10 mini-gels (10 × 10 cm) or 5 gels or membranes (16.5 × 19 cm)