MilliporeSigma
  • Heteromerization of the gammac chain with the interleukin-9 receptor alpha subunit leads to STAT activation and prevention of apoptosis.
Heteromerization of the gammac chain with the interleukin-9 receptor alpha subunit leads to STAT activation and prevention of apoptosis.
The Journal of biological chemistry (1998-05-16)
J H Bauer, K D Liu, Y You, S Y Lai, M A Goldsmith
ABSTRACT
Interleukin-9 (IL-9) is a cytokine with pleiotropic effects on mast cell and T cell lines. It exerts its effects through the IL-9R complex consisting of IL-9Ralpha and the common gammac subunit. Here we report functional evidence for receptor heteromerization for efficient signal transduction, and we define minimal requirements in the two receptor subunits for IL-9R function. Tyrosine 336 of the IL-9Ralpha and the membrane-proximal segment of gammac are both crucial for signaling. The activated IL-9R complex employs the Janus kinases JAK1 and JAK3 for subsequent activation of the signal transducer and activator transcription (STAT) factors STAT-1, STAT-3, and STAT-5. This process is independent of Tyk2. We demonstrate further that the activated STAT complexes consist of STAT-1 and STAT-5 homodimers and STAT-1-STAT-3 heterodimers. Finally, we show that IL-9R signaling in a T cell line does not result in detectable mitogen-activated protein kinase activation and leads to unsustained proliferation. Nonetheless, these T cells are efficiently protected from dexamethasone-induced apoptosis. These results further define the molecular architecture of the IL-9R and its specific connections to various biologic responses.