MilliporeSigma
  • Identification and cloning of a glucan- and lipopolysaccharide-binding protein from Eisenia foetida earthworm involved in the activation of prophenoloxidase cascade.
Identification and cloning of a glucan- and lipopolysaccharide-binding protein from Eisenia foetida earthworm involved in the activation of prophenoloxidase cascade.
The Journal of biological chemistry (1998-09-12)
A Beschin, M Bilej, F Hanssens, J Raymakers, E Van Dyck, H Revets, L Brys, J Gomez, P De Baetselier, M Timmermans
ABSTRACT
Coelomic fluid of Eisenia foetida earthworms contains a 42-kDa protein named coelomic cytolytic factor 1 (CCF-1) that was described previously to be involved in cytolytic, opsonizing, and hemolytic properties of the coelomic fluid. Cloning and sequencing of CCF-1 reveal significant homology with the putative catalytic region of beta-1,3- and beta-1,3-1,4-glucanases. CCF-1 also displays homology with coagulation factor G from Limulus polyphemus and with Gram-negative bacteria-binding protein of Bombyx mori silkworm, two proteins involved in invertebrate defense mechanisms. We show that CCF-1 efficiently binds both beta-1,3-glucan and lipopolysaccharide. Moreover, CCF-1 participates in the activation of prophenoloxidase cascade via recognition of yeast and Gram-negative bacteria cell wall components. These results suggest that the 42-kDa CCF-1 protein of E. foetida coelomic fluid likely plays a role in the protection of earthworms against microbes.
Sigma-Aldrich
β-Gentiobiose, ≥85% (remainder primarily α-anomer)