MilliporeSigma
  • A 37 kb region upstream of brachyury comprising a notochord enhancer is essential for notochord and tail development.
A 37 kb region upstream of brachyury comprising a notochord enhancer is essential for notochord and tail development.
Development (Cambridge, England) (2021-11-26)
Dennis Schifferl, Manuela Scholze-Wittler, Lars Wittler, Jesse V Veenvliet, Frederic Koch, Bernhard G Herrmann
ABSTRACT
The node-streak border region comprising notochord progenitor cells (NPCs) at the posterior node and neuro-mesodermal progenitor cells (NMPs) in the adjacent epiblast is the prime organizing center for axial elongation in mouse embryos. The T-box transcription factor brachyury (T) is essential for both formation of the notochord and maintenance of NMPs, and thus is a key regulator of trunk and tail development. The T promoter controlling T expression in NMPs and nascent mesoderm has been characterized in detail; however, control elements for T expression in the notochord have not been identified yet. We have generated a series of deletion alleles by CRISPR/Cas9 genome editing in mESCs, and analyzed their effects in mutant mouse embryos. We identified a 37 kb region upstream of T that is essential for notochord function and tailbud outgrowth. Within that region, we discovered a T-binding enhancer required for notochord cell specification and differentiation. Our data reveal a complex regulatory landscape controlling cell type-specific expression and function of T in NMP/nascent mesoderm and node/notochord, allowing proper trunk and tail development.
Sigma-Aldrich
Agarose, low gelling temperature, BioReagent, for molecular biology
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
Histodenz, nonionic density gradient medium
Roche
Hygromycin B, from Streptomyces hygroscopicus
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline