MilliporeSigma
  • G-quadruplexes are transcription factor binding hubs in human chromatin.
G-quadruplexes are transcription factor binding hubs in human chromatin.
Genome biology (2021-04-25)
Jochen Spiegel, Sergio Martínez Cuesta, Santosh Adhikari, Robert Hänsel-Hertsch, David Tannahill, Shankar Balasubramanian
ABSTRACT
The binding of transcription factors (TF) to genomic targets is critical in the regulation of gene expression. Short, double-stranded DNA sequence motifs are routinely implicated in TF recruitment, but many questions remain on how binding site specificity is governed. Herein, we reveal a previously unappreciated role for DNA secondary structures as key features for TF recruitment. In a systematic, genome-wide study, we discover that endogenous G-quadruplex secondary structures (G4s) are prevalent TF binding sites in human chromatin. Certain TFs bind G4s with affinities comparable to double-stranded DNA targets. We demonstrate that, in a chromatin context, this binding interaction is competed out with a small molecule. Notably, endogenous G4s are prominent binding sites for a large number of TFs, particularly at promoters of highly expressed genes. Our results reveal a novel non-canonical mechanism for TF binding whereby G4s operate as common binding hubs for many different TFs to promote increased transcription.
Sigma-Aldrich
Anti-CEBPZ antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Anti-CTCF Antibody, serum, Upstate®