MilliporeSigma
  • Endoplasmic reticulum acetyltransferases Atase1 and Atase2 differentially regulate reticulophagy, macroautophagy and cellular acetyl-CoA metabolism.
Endoplasmic reticulum acetyltransferases Atase1 and Atase2 differentially regulate reticulophagy, macroautophagy and cellular acetyl-CoA metabolism.
Communications biology (2021-04-14)
Michael J Rigby, Alexis J Lawton, Gulpreet Kaur, Varuna C Banduseela, William E Kamm, Aparna Lakkaraju, John M Denu, Luigi Puglielli
ABSTRACT
Nε-lysine acetylation in the ER lumen is a recently discovered quality control mechanism that ensures proteostasis within the secretory pathway. The acetyltransferase reaction is carried out by two type-II membrane proteins, ATase1/NAT8B and ATase2/NAT8. Prior studies have shown that reducing ER acetylation can induce reticulophagy, increase ER turnover, and alleviate proteotoxic states. Here, we report the generation of Atase1-/- and Atase2-/- mice and show that these two ER-based acetyltransferases play different roles in the regulation of reticulophagy and macroautophagy. Importantly, knockout of Atase1 alone results in activation of reticulophagy and rescue of the proteotoxic state associated with Alzheimer's disease. Furthermore, loss of Atase1 or Atase2 results in widespread adaptive changes in the cell acetylome and acetyl-CoA metabolism. Overall, our study supports a divergent role of Atase1 and Atase2 in cellular biology, emphasizing ATase1 as a valid translational target for diseases characterized by toxic protein aggregation in the secretory pathway.
Sigma-Aldrich
Anti-ATF6 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Monoclonal Anti-Actin, Smooth Muscle antibody produced in mouse, ~1.0 mg/mL, clone CGA7, purified immunoglobulin, buffered aqueous solution