MilliporeSigma
  • Chromatin lncRNA Platr10 controls stem cell pluripotency by coordinating an intrachromosomal regulatory network.
Chromatin lncRNA Platr10 controls stem cell pluripotency by coordinating an intrachromosomal regulatory network.
Genome biology (2021-08-21)
Zhonghua Du, Xue Wen, Yichen Wang, Lin Jia, Shilin Zhang, Yudi Liu, Lei Zhou, Hui Li, Wang Yang, Cong Wang, Jingcheng Chen, Yajing Hao, Daniela Salgado Figueroa, Huiling Chen, Dan Li, Naifei Chen, Ilkay Celik, Yanbo Zhu, Zi Yan, Changhao Fu, Shanshan Liu, Benzheng Jiao, Zhuo Wang, Hui Zhang, Günhan Gülsoy, Jianjun Luo, Baoming Qin, Sujun Gao, Philipp Kapranov, Miguel A Esteban, Songling Zhang, Wei Li, Ferhat Ay, Runsheng Chen, Andrew R Hoffman, Jiuwei Cui, Ji-Fan Hu
ABSTRACT
A specific 3-dimensional intrachromosomal architecture of core stem cell factor genes is required to reprogram a somatic cell into pluripotency. As little is known about the epigenetic readers that orchestrate this architectural remodeling, we used a novel chromatin RNA in situ reverse transcription sequencing (CRIST-seq) approach to profile long noncoding RNAs (lncRNAs) in the Oct4 promoter. We identify Platr10 as an Oct4 - Sox2 binding lncRNA that is activated in somatic cell reprogramming. Platr10 is essential for the maintenance of pluripotency, and lack of this lncRNA causes stem cells to exit from pluripotency. In fibroblasts, ectopically expressed Platr10 functions in trans to activate core stem cell factor genes and enhance pluripotent reprogramming. Using RNA reverse transcription-associated trap sequencing (RAT-seq), we show that Platr10 interacts with multiple pluripotency-associated genes, including Oct4, Sox2, Klf4, and c-Myc, which have been extensively used to reprogram somatic cells. Mechanistically, we demonstrate that Platr10 helps orchestrate intrachromosomal promoter-enhancer looping and recruits TET1, the enzyme that actively induces DNA demethylation for the initiation of pluripotency. We further show that Platr10 contains an Oct4 binding element that interacts with the Oct4 promoter and a TET1-binding element that recruits TET1. Mutation of either of these two elements abolishes Platr10 activity. These data suggest that Platr10 functions as a novel chromatin RNA molecule to control pluripotency in trans by modulating chromatin architecture and regulating DNA methylation in the core stem cell factor network.
Product Description
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Fluorescent Mouse ES/iPS Cell Characterization Kit, This Fluorescent Mouse ES/iPS Cell Characterization Kit contains a range of sensitive tools for the phenotypic assessment of the pluripotent status of mouse Embryonic stem & induced pluripotent Stem cells.