MilliporeSigma
Site-Specific Protein Labeling via Sortase-Mediated Transpeptidation.
Current protocols in protein science (2017-08-02)
John M Antos, Jessica Ingram, Tao Fang, Novalia Pishesha, Matthias C Truttmann, Hidde L Ploegh
ABSTRACT
Strategies for site-specific protein modification are highly desirable for the construction of conjugates containing non-genetically-encoded functional groups. Ideally, these strategies should proceed under mild conditions, and be compatible with a wide range of protein targets and non-natural moieties. The transpeptidation reaction catalyzed by bacterial sortases is a prominent strategy for protein derivatization that possesses these features. Naturally occurring or engineered variants of sortase A from Staphylococcus aureus catalyze a ligation reaction between a five-amino-acid substrate motif (LPXTG) and oligoglycine nucleophiles. By pairing proteins and synthetic peptides that possess these ligation handles, it is possible to install modifications onto the protein N- or C-terminus in site-specific fashion. As described in this unit, the successful implementation of sortase-mediated labeling involves straightforward solid-phase synthesis and molecular biology techniques, and this method is compatible with proteins in solution or on the surface of live cells. © 2017 by John Wiley & Sons, Inc.
Sigma-Aldrich
Kaiser test kit