MilliporeSigma
  • Limited TCR repertoire and ENTPD1 dysregulation mark late-stage COVID-19.
Limited TCR repertoire and ENTPD1 dysregulation mark late-stage COVID-19.
iScience (2021-10-06)
Na Wang, Marta Vuerich, Ahmadreza Kalbasi, Jonathon J Graham, Eva Csizmadia, Zachary James Manickas-Hill, Ann Woolley, Clement David, Eric M Miller, Kara Gorman, Jonathan L Hecht, Shahzad Shaefi, Simon C Robson, Maria Serena Longhi
ABSTRACT
T cell exhaustion and dysfunction are hallmarks of severe COVID-19. To gain insights into the pathways underlying these alterations, we performed a comprehensive transcriptome analysis of peripheral-blood-mononuclear-cells (PBMCs), spleen, lung, kidney, liver, and heart obtained at autopsy from COVID-19 patients and matched controls, using the nCounter CAR-T-Characterization panel. We found substantial gene alterations in COVID-19-impacted organs, especially the lung where altered TCR repertoires are noted. Reduced TCR repertoires are also observed in PBMCs of severe COVID-19 patients. ENTPD1/CD39, an ectoenzyme defining exhausted T-cells, is upregulated in the lung, liver, spleen, and PBMCs of severe COVID-19 patients where expression positively correlates with markers of vasculopathy. Heightened ENTPD1/CD39 is paralleled by elevations in STAT-3 and HIF-1α transcription factors; and by markedly reduced CD39-antisense-RNA, a long-noncoding-RNA negatively regulating ENTPD1/CD39 at the post-transcriptional level. Limited TCR repertoire and aberrant regulation of ENTPD1/CD39 could have permissive roles in COVID-19 progression and indicate potential therapeutic targets to reverse disease.
Sigma-Aldrich
Anti-ENTPD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution