MilliporeSigma
  • Glycogen Phosphorylase Isoform Regulation of Ventromedial Hypothalamic Nucleus Gluco-Regulatory Neuron 5'-AMP-Activated Protein Kinase and Transmitter Marker Protein Expression.
Glycogen Phosphorylase Isoform Regulation of Ventromedial Hypothalamic Nucleus Gluco-Regulatory Neuron 5'-AMP-Activated Protein Kinase and Transmitter Marker Protein Expression.
ASN neuro (2021-10-02)
Md Main Uddin, Mostafa M H Ibrahim, Karen P Briski
ABSTRACT
Brain glycogen is remodeled during metabolic homeostasis and provides oxidizable L-lactate equivalents. Brain glycogen phosphorylase (GP)-brain (GPbb; AMP-sensitive) and -muscle (GPmm; norepinephrine-sensitive) type isoforms facilitate stimulus-specific control of glycogen disassembly. Here, a whole animal model involving stereotactic-targeted delivery of GPmm or GPbb siRNA to the ventromedial hypothalamic nucleus (VMN) was used to investigate the premise that these variants impose differential control of gluco-regulatory transmission. Intra-VMN GPmm or GPbb siRNA administration inhibited glutamate decarboxylate65/67 (GAD), a protein marker for the gluco-inhibitory transmitter γ--aminobutyric acid (GABA), in the caudal VMN. GPbb knockdown, respectively overturned or exacerbated hypoglycemia-associated GAD suppression in rostral and caudal VMN. GPmm siRNA caused a segment-specific reversal of hypoglycemic augmentation of the gluco-stimulatory transmitter indicator, neuronal nitric oxide synthase (nNOS). In both cell types, GP siRNA down-regulated 5'-AMP-activated protein kinase (AMPK) during euglycemia, but hypoglycemic suppression of AMPK was reversed by GPmm targeting. GP knockdown elevated baseline GABA neuron phosphoAMPK (pAMKP) content, and amplified hypoglycemic augmentation of pAMPK expression in each neuron type. GPbb knockdown increased corticosterone secretion in eu- and hypoglycemic rats. Outcomes validate efficacy of GP siRNA delivery for manipulation of glycogen breakdown in discrete brain structures in vivo, and document VMN GPbb control of local GPmm expression. Results document GPmm and/or -bb regulation of GABAergic and nitrergic transmission in discrete rostro-caudal VMN segments. Contrary effects of glycogenolysis on metabolic-sensory AMPK protein during eu- versus hypoglycemia may reflect energy state-specific astrocyte signaling. Amplifying effects of GPbb knockdown on hypoglycemic stimulation of pAMPK infer that glycogen mobilization by GPbb limits neuronal energy instability during hypoglycemia.
Sigma-Aldrich
Anti-GAD 65/67, from rabbit, purified by affinity chromatography