MilliporeSigma
  • Effects of long-term social isolation on central, behavioural and metabolic parameters in middle-aged mice.
Effects of long-term social isolation on central, behavioural and metabolic parameters in middle-aged mice.
Behavioural brain research (2021-10-18)
Izabelle Dias Benfato, Ana Carolina Silvares Quintanilha, Jessica Salles Henrique, Melyssa Alves Souza, Barbara Dos Anjos Rosário, Jose Ivo Araújo Beserra Filho, Robson Luiz Oliveira Santos, Alessandra Mussi Ribeiro, Luciana Le Sueur Maluf, Camila Aparecida Machado de Oliveira
ABSTRACT
Social isolation gained discussion momentum due to the COVID-19 pandemic. Whereas many studies address the effects of long-term social isolation in post-weaning and adolescence and for periods ranging from 4 to 12 weeks, little is known about the repercussions of adult long-term social isolation in middle age. Thus, our aim was to investigate how long-term social isolation can influence metabolic, behavioural, and central nervous system-related areas in middle-aged mice. Adult male C57Bl/6 mice (4 months-old) were randomly divided into Social (2 cages, n = 5/cage) and Isolated (10 cages, n = 1/cage) housing groups, totalizing 30 weeks of social isolation, which ended concomitantly with the onset of middle age of mice. At the end of the trial, metabolic parameters, short-term memory, anxiety-like behaviour, and physical activity were assessed. Immunohistochemistry in the hippocampus (ΔFosB, BDNF, and 8OHDG) and hypothalamus (ΔFosB) was also performed. The Isolated group showed impaired memory along with a decrease in hippocampal ΔFosB at dentate gyrus and in BDNF at CA3. Food intake was also affected, but the direction depended on how it was measured in the Social group (individually or in the group) with no alteration in ΔFosB at the hypothalamus. Physical activity parameters increased with chronic isolation, but in the light cycle (inactive phase), with some evidence of anxiety-like behaviour. Future studies should better explore the timepoint at which the alterations found begin. In conclusion, long-term social isolation in adult mice contributes to alterations in feeding, physical activity pattern, and anxiety-like behaviour. Moreover, short-term memory deficit was associated with lower levels of hippocampal ΔFosB and BDNF in middle age.
Sigma-Aldrich
Anti-FOSB antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody