MilliporeSigma
  • Immunochemical properties of a 60 kDa cell surface-associated heat shock-protein (Hsp60) from Helicobacter pylori.
Immunochemical properties of a 60 kDa cell surface-associated heat shock-protein (Hsp60) from Helicobacter pylori.
FEMS immunology and medical microbiology (1996-12-31)
H R Amini, F Ascencio, A Cruz-Villacorta, E Ruiz-Bustos, T Wadström
ABSTRACT
Western blot analysis (immunoblotting) of cell surface-associated proteins from Helicobacter pylori confirmed our previous findings that binding of human IgG is a common property (among H. pylori strains). Purification of the IgG-binding proteins (IGBP) was achieved by two purification steps, affinity chromatography on IgG-Sepharose and nickel chelate affinity chromatography. SDS-PAGE and immunoblotting analysis revealed a 60 kDa protein with affinity for peroxidase labeled human IgG. Solid phase binding assays showed that IgG binds to an immobilized protein (IGBP). The 60 kDa IGBP binds human IgG1, IgG3 and IgM. Binding could be inhibited by the kappa chain of the human IgG, but not with its Fc fragment, nor with IgA or IgM. In addition, rabbit polyclonal antibodies raised against the 60 kDa IGBP blocked IgG binding. Monoclonal antibodies, specific to the Hsp60 heat shock protein of H. pylori recognized the 60 kDa IGBP as revealed by immunoblotting analysis, both in crude preparations and in the purified fractions.
Sigma-Aldrich
IgM from human serum, reagent grade, ~95% (HPLC), buffered aqueous solution