MilliporeSigma
  • NRF3 upregulates gene expression in SREBP2-dependent mevalonate pathway with cholesterol uptake and lipogenesis inhibition.
NRF3 upregulates gene expression in SREBP2-dependent mevalonate pathway with cholesterol uptake and lipogenesis inhibition.
iScience (2021-10-21)
Tsuyoshi Waku, Toru Hagiwara, Natsuko Tamura, Yuri Atsumi, Yasuomi Urano, Mikiko Suzuki, Takuya Iwami, Katsuya Sato, Masayuki Yamamoto, Noriko Noguchi, Akira Kobayashi
ABSTRACT
Lipids, such as cholesterol and fatty acids, influence cell signaling, energy storage, and membrane formation. Cholesterol is biosynthesized through the mevalonate pathway, and aberrant metabolism causes metabolic diseases. The genetic association of a transcription factor NRF3 with obesity has been suggested, although the molecular mechanisms remain unknown. Here, we show that NRF3 upregulates gene expression in SREBP2-dependent mevalonate pathway. We further reveal that NRF3 overexpression not only reduces lanosterol, a cholesterol precursor, but also induces the expression of the GGPS1 gene encoding an enzyme in the production of GGPP from farnesyl pyrophosphate (FPP), a lanosterol precursor. NRF3 overexpression also enhances cholesterol uptake through RAB5-mediated macropinocytosis process, a bulk and fluid-phase endocytosis pathway. Moreover, we find that GGPP treatment abolishes NRF3 knockdown-mediated increase of neutral lipids. These results reveal the potential roles of NRF3 in the SREBP2-dependent mevalonate pathway for cholesterol uptake through macropinocytosis induction and for lipogenesis inhibition through GGPP production.
Sigma-Aldrich
Nile Red
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-Farnesyl Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-HMG-CoA Reductase Antibody, clone IgG-A9, clone IgG-A9, from mouse