MilliporeSigma
  • Synthesis, photoreactivity and cytotoxic activity of caged compounds of L-leucyl-L-leucine methyl ester, an apoptosis inducer.
Synthesis, photoreactivity and cytotoxic activity of caged compounds of L-leucyl-L-leucine methyl ester, an apoptosis inducer.
Photochemistry and photobiology (1996-06-01)
M Odaka, T Furuta, Y Kobayashi, M Iwamura
ABSTRACT
L-Leucyl-L-leucine methyl ester (Leu-Leu-OMe), an apoptosis inducer in natural killer cells and macrophages, was caged with trans-o-hydroxycinnamoyl (3a-d), trans-o-mercaptocinnamoyl (4) and o-nitrobenzyl derivatives (5a, b), and the photochemical reactivity of these derivatives in phosphate-buffered saline containing 1% dimethyl sulfoxide and their immunological properties were studied. All of the derivatives exhibited absorption at wavelengths longer than the UVB region. Although 3a-d and 4 were expected to isomerize to a cis isomer, which then cyclizes intramolecularly to give Leu-Leu-OMe and a coumarin derivative, cyclization efficiency was not satisfactory except for 3a. However, 3a itself caused necrosis (cell swelling) of U937 cells (a myeloid cell line). In contrast, 5a and b released Leu-Le-OMe quickly and efficiently and did not affect U937 cells. Although irradiated 5b induced necrosis, irradiated 3a and 5a induced apoptosis in these cells, as evidenced by a decrease in cell size.
Sigma-Aldrich
Leu-Leu methyl ester hydrobromide, ≥97% (TLC)