MilliporeSigma
  • Nucleolin stabilizes G-quadruplex structures folded by the LTR promoter and silences HIV-1 viral transcription.
Nucleolin stabilizes G-quadruplex structures folded by the LTR promoter and silences HIV-1 viral transcription.
Nucleic acids research (2015-09-12)
Elena Tosoni, Ilaria Frasson, Matteo Scalabrin, Rosalba Perrone, Elena Butovskaya, Matteo Nadai, Giorgio Palù, Dan Fabris, Sara N Richter
ABSTRACT
Folding of the LTR promoter into dynamic G-quadruplex conformations has been shown to suppress its transcriptional activity in HIV-1. Here we sought to identify the proteins that control the folding of this region of proviral genome by inducing/stabilizing G-quadruplex structures. The implementation of electrophorethic mobility shift assay and pull-down experiments coupled with mass spectrometric analysis revealed that the cellular protein nucleolin is able to specifically recognize G-quadruplex structures present in the LTR promoter. Nucleolin recognized with high affinity and specificity the majority, but not all the possible G-quadruplexes folded by this sequence. In addition, it displayed greater binding preference towards DNA than RNA G-quadruplexes, thus indicating two levels of selectivity based on the sequence and nature of the target. The interaction translated into stabilization of the LTR G-quadruplexes and increased promoter silencing activity; in contrast, disruption of nucleolin binding in cells by both siRNAs and a nucleolin binding aptamer greatly increased LTR promoter activity. These data indicate that nucleolin possesses a specific and regulated activity toward the HIV-1 LTR promoter, which is mediated by G-quadruplexes. These observations provide new essential insights into viral transcription and a possible low mutagenic target for antiretroviral therapy.
Sigma-Aldrich
NuCLEAR Extraction Kit, For mammalian tissue or cultured cells