MilliporeSigma
  • Proteomic study reveals downregulation of apolipoprotein A1 in plasma of poorly controlled diabetes: a pilot study.
Proteomic study reveals downregulation of apolipoprotein A1 in plasma of poorly controlled diabetes: a pilot study.
Molecular medicine reports (2012-12-13)
Hemangi S Bhonsle, Arvind M Korwar, Ashok D Chougale, Sachin S Kote, Nitin L Dhande, Kishore M Shelgikar, Mahesh J Kulkarni
ABSTRACT
Proteomic approaches aid in gaining a better understanding of the pathophysiology of diabetic complications. In view of this, differential protein expression in diabetic plasma samples was studied by a combination of proteomic and western blot analyses. Diabetic plasma samples were categorized based on glycated haemoglobin levels as controlled diabetes (CD; 7-8%), poorly controlled diabetes (PCD; >8%) and non-diabetic control (ND;<6.4%). Two-dimensional electrophoresis and liquid chromatography‑mass spectrometry revealed differential expression of proteins including upregulation of fibrinogen and haptoglobin and downregulation of vitamin D binding protein, α-1-antitrypsin, transthyretin and apolipoprotein A1 (Apo A1) in diabetic compared with non-diabetic plasma samples. Amongst these proteins, Apo A1 downregulation was prominent in PCD. Downregulation of Apo A1 may serve as an early predictive marker of diabetic complications.
Product Description
Millipore
ProteoPrep® Blue Albumin & IgG Depletion Kit