MilliporeSigma
  • Transcriptionally inactive hepatitis B virus episome DNA preferentially resides in the vicinity of chromosome 19 in 3D host genome upon infection.
Transcriptionally inactive hepatitis B virus episome DNA preferentially resides in the vicinity of chromosome 19 in 3D host genome upon infection.
Cell reports (2021-07-01)
Dingbin Tang, Hanqing Zhao, Yumeng Wu, Bo Peng, Zhenchao Gao, Yinyan Sun, Jinzhi Duan, Yonghe Qi, Yunfei Li, Zhongmin Zhou, Guilan Guo, Yu Zhang, Cheng Li, Jianhua Sui, Wenhui Li
ABSTRACT
The hepatitis B virus (HBV) infects 257 million people worldwide. HBV infection requires establishment and persistence of covalently closed circular (ccc) DNA, a viral episome, in nucleus. Here, we study cccDNA spatial localization in the 3D host genome by using chromosome conformation capture-based sequencing analysis and fluorescence in situ hybridization (FISH). We show that transcriptionally inactive cccDNA is not randomly distributed in host nucleus. Rather, it is preferentially accumulated at specialized areas, including regions close to chromosome 19 (chr.19). Activation of the cccDNA is apparently associated with its re-localization, from a pre-established heterochromatin hub formed by 5 regions of chr.19 to transcriptionally active regions formed by chr.19 and nearby chromosomes including chr.16, 17, 20, and 22. This active versus inactive positioning at discrete regions of the host genome is primarily controlled by the viral HBx protein and by host factors including the structural maintenance of chromosomes protein 5/6 (SMC5/6) complex.
Sigma-Aldrich
Anti-SMC6 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution