MilliporeSigma
  • De novo identification of mammalian ciliary motility proteins using cryo-EM.
De novo identification of mammalian ciliary motility proteins using cryo-EM.
Cell (2021-10-30)
Miao Gui, Hannah Farley, Priyanka Anujan, Jacob R Anderson, Dale W Maxwell, Jonathan B Whitchurch, J Josephine Botsch, Tao Qiu, Shimi Meleppattu, Sandeep K Singh, Qi Zhang, James Thompson, Jane S Lucas, Colin D Bingle, Dominic P Norris, Sudipto Roy, Alan Brown
ABSTRACT
Dynein-decorated doublet microtubules (DMTs) are critical components of the oscillatory molecular machine of cilia, the axoneme, and have luminal surfaces patterned periodically by microtubule inner proteins (MIPs). Here we present an atomic model of the 48-nm repeat of a mammalian DMT, derived from a cryoelectron microscopy (cryo-EM) map of the complex isolated from bovine respiratory cilia. The structure uncovers principles of doublet microtubule organization and features specific to vertebrate cilia, including previously unknown MIPs, a luminal bundle of tektin filaments, and a pentameric dynein-docking complex. We identify a mechanism for bridging 48- to 24-nm periodicity across the microtubule wall and show that loss of the proteins involved causes defective ciliary motility and laterality abnormalities in zebrafish and mice. Our structure identifies candidate genes for diagnosis of ciliopathies and provides a framework to understand their functions in driving ciliary motility.
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Adenosine 5′-triphosphate magnesium salt, ≥95%, bacterial
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in glass vials
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Roche
DIG RNA Labeling Mix, sufficient for 20 reactions, solution
Roche
INT/BCIP Stock Solution, solution, pkg of 3 mL