MilliporeSigma
  • Identification of theranostic factors for patients developing metastasis after surgery for early-stage lung adenocarcinoma.
Identification of theranostic factors for patients developing metastasis after surgery for early-stage lung adenocarcinoma.
Theranostics (2021-03-06)
Wei-Chung Cheng, Chun-Yu Chang, Chia-Chien Lo, Chih-Ying Hsieh, Ting-Ting Kuo, Guan-Chin Tseng, Sze-Ching Wong, Shu-Fen Chiang, Kevin Chih-Yang Huang, Liang-Chuan Lai, Tzu-Pin Lu, K S Clifford Chao, Yuh-Pyng Sher
ABSTRACT
Rationale: Lung adenocarcinoma (LUAD) is an aggressive disease with high propensity of metastasis. Among patients with early-stage disease, more than 30% of them may relapse or develop metastasis. There is an unmet medical need to stratify patients with early-stage LUAD according to their risk of relapse/metastasis to guide preventive or therapeutic approaches. In this study, we identified 4 genes that can serve both therapeutic and diagnostic (theranostic) purposes. Methods: Three independent datasets (GEO, TCGA, and KMPlotter) were used to evaluate gene expression profile of patients with LUAD by unbiased screening approach. Upon significant genes uncovered, functional enrichment analysis was carried out. The predictive power of their expression on patient prognosis were evaluated. Once confirmed their theranostic roles by integrated bioinformatics, we further conducted in vitro and in vivo validation. Results: We found that four genes (ADAM9, MTHFD2, RRM2, and SLC2A1) were associated with poor patient outcomes with an increased hazard ratio in LUAD. Knockdown of them, both separately and simultaneously, suppressed lung cancer cell proliferation and migration ability in vitro and prolonged survival time in metastatic tumor mouse models. Moreover, these four biomarkers were found to be overexpressed in tumor tissues from LUAD patients, and the total immunohistochemical staining scores correlated with poor prognosis. Conclusions: These results suggest that these four identified genes could be theranostic biomarkers for stratifying high-risk patients who develop relapse/metastasis in early-stage LUAD. Developing therapeutic approaches for the four biomarkers may benefit early-stage LUAD patients after surgery.
Sigma-Aldrich
Anti-RRM2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution