MilliporeSigma
  • Microtubule dynamics influence the retrograde biased motility of kinesin-4 motor teams in neuronal dendrites.
Microtubule dynamics influence the retrograde biased motility of kinesin-4 motor teams in neuronal dendrites.
Molecular biology of the cell (2021-10-28)
Erin M Masucci, Peter K Relich, Melike Lakadamyali, E Michael Ostap, Erika L F Holzbaur
ABSTRACT
Microtubules establish the directionality of intracellular transport by kinesins and dynein through polarized assembly, but it remains unclear how directed transport occurs along microtubules organized with mixed polarity. We investigated the ability of the plus-end directed kinesin-4 motor KIF21B to navigate mixed polarity microtubules in mammalian dendrites. Reconstitution assays with recombinant KIF21B and engineered microtubule bundles or extracted neuronal cytoskeletons indicate that nucleotide-independent microtubule binding regions of KIF21B modulate microtubule dynamics and promote directional switching on antiparallel microtubules. Optogenetic recruitment of KIF21B to organelles in live neurons induces unidirectional transport in axons but bi-directional transport with a net retrograde bias in dendrites. Removal of the secondary microtubule binding regions of KIF21B or dampening of microtubule dynamics with low concentrations of nocodazole eliminates retrograde bias in live dendrites. Further exploration of the contribution of microtubule dynamics in dendrites to directionality revealed plus-end-out microtubules to be more dynamic than plus-end-in microtubules, with nocodazole preferentially stabilizing the plus-end-out population. We propose a model in which both nucleotide-sensitive and insensitive microtubule binding sites of KIF21B motors contribute to the search and selection of stable plus-end-in microtubules within the mixed polarity microtubule arrays characteristic of mammalian dendrites to achieve net retrograde movement of KIF21B-bound cargos. [Media: see text] [Media: see text] [Media: see text] [Media: see text] [Media: see text] [Media: see text] [Media: see text].
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-MAP2 Antibody, clone AP20, clone AP20, Chemicon®, from mouse
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
Sigma-Aldrich
Anti-MAP1 Antibody, clone HM-1, ascites fluid, clone HM-1, Chemicon®
Sigma-Aldrich
Anti-Tubulin Antibody, clone YL1/2, clone YL1/2, Chemicon®, from rat
Sigma-Aldrich
Anti-Beta III Tubulin Antibody, Chemicon®, from chicken