MilliporeSigma
  • Elevated Cholesteryl Ester Transfer Protein Activity Early in Pregnancy Predicts Prediabetes 5 Years Later.
Elevated Cholesteryl Ester Transfer Protein Activity Early in Pregnancy Predicts Prediabetes 5 Years Later.
The Journal of clinical endocrinology and metabolism (2019-10-31)
Thor Ueland, Marie Cecilie Paasche Roland, Annika E Michelsen, Kristin Godang, Pål Aukrust, Tore Henriksen, Jens Bollerslev, Tove Lekva
ABSTRACT
Cholesteryl ester transfer protein (CETP) regulates high-density lipoprotein (HDL) cholesterol levels and interaction between glucose, and HDL metabolism is central in the development of diabetes. We hypothesized that CETP levels would be regulated in diabetic pregnancies. We tested the hypothesis by evaluating CETP activity measured multiple times during pregnancy and at 5 years' follow-up in a prospective cohort (STORK) and investigated its association with gestational diabetes mellitus (GDM) during pregnancy or development of prediabetes 5 years after pregnancy. We also evaluated the strongest correlation of CETP activity among measures of adipocity and glucose metabolism, lipoproteins, adipokines, and monocyte/macrophage activation markers. A population-based longitudinal cohort study was conducted from 2001 to 2013. The study setting was Oslo University Hospital. A total of 300 women during pregnancy and at 5 years postpartum participated in this study. CETP activity was measured at 14 to 16, 22 to 24, 30 to 32, and 36 to 38 weeks' gestation, and at 5 years' follow-up. We found higher CETP activity in pregnancy in women developing prediabetes but no association with GDM. CETP activity decreased throughout pregnancy and remained low at follow-up. High CETP activity was associated with sCD14 levels, in particular in women who developed prediabetes. These data show that enhanced CETP activity during pregnancy is associated with systemic indices of monocyte/macrophage activation, in particular in women who develop prediabetes later in life. CETP activity during pregnancy identifies women at risk for later diabetes development.
Sigma-Aldrich
CETP Activity Assay Kit, Supplied by Roar Biomedical, Inc.