MilliporeSigma
  • USP42 protects ZNRF3/RNF43 from R-spondin-dependent clearance and inhibits Wnt signalling.
USP42 protects ZNRF3/RNF43 from R-spondin-dependent clearance and inhibits Wnt signalling.
EMBO reports (2021-04-01)
Nicole Giebel, Anchel de Jaime-Soguero, Ana García Del Arco, Jonathan J M Landry, Marlene Tietje, Laura Villacorta, Vladimir Benes, Vanesa Fernández-Sáiz, Sergio P Acebrón
ABSTRACT
The tumour suppressors RNF43 and ZNRF3 play a central role in development and tissue homeostasis by promoting the turnover of the Wnt receptors LRP6 and Frizzled (FZD). The stem cell growth factor R-spondin induces auto-ubiquitination and membrane clearance of ZNRF3/RNF43 to promote Wnt signalling. However, the deubiquitinase stabilising ZNRF3/RNF43 at the plasma membrane remains unknown. Here, we show that the USP42 antagonises R-spondin by protecting ZNRF3/RNF43 from ubiquitin-dependent clearance. USP42 binds to the Dishevelled interacting region (DIR) of ZNRF3 and stalls the R-spondin-LGR4-ZNRF3 ternary complex by deubiquitinating ZNRF3. Accordingly, USP42 increases the turnover of LRP6 and Frizzled (FZD) receptors and inhibits Wnt signalling. Furthermore, we show that USP42 functions as a roadblock for paracrine Wnt signalling in colon cancer cells and mouse small intestinal organoids. We provide new mechanistic insights into the regulation R-spondin and conclude that USP42 is crucial for ZNRF3/RNF43 stabilisation at the cell surface.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Anti-USP42 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid