MilliporeSigma
  • miR-1 sustains muscle physiology by controlling V-ATPase complex assembly.
miR-1 sustains muscle physiology by controlling V-ATPase complex assembly.
Science advances (2021-10-16)
Paula Gutiérrez-Pérez, Emilio M Santillán, Thomas Lendl, Jingkui Wang, Anna Schrempf, Thomas L Steinacker, Mila Asparuhova, Marlene Brandstetter, David Haselbach, Luisa Cochella
ABSTRACT
[Figure: see text].
Sigma-Aldrich
Protease from Streptomyces griseus, BioReagent, DNase, RNase, and nickase, none detected (No RNase.)
Sigma-Aldrich
Penicillin-Streptomycin, Solution Stabilized, with 5,000 units penicillin and 5mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Chitinase from Streptomyces griseus, lyophilized powder (essentially salt free), ≥200 units/g solid