MilliporeSigma
  • NAD+ supplement potentiates tumor-killing function by rescuing defective TUB-mediated NAMPT transcription in tumor-infiltrated T cells.
NAD+ supplement potentiates tumor-killing function by rescuing defective TUB-mediated NAMPT transcription in tumor-infiltrated T cells.
Cell reports (2021-08-12)
Yuetong Wang, Fei Wang, Lihua Wang, Shizhen Qiu, Yufeng Yao, Chenxu Yan, Xuexue Xiong, Xuyong Chen, Quanquan Ji, Jian Cao, Ganglong Gao, Dake Li, Liye Zhang, Zhiqian Guo, Ruoning Wang, Haopeng Wang, Gaofeng Fan
ABSTRACT
Although tumor-infiltrating lymphocytes (TILs) maintain their ability to proliferate, persist, and eradicate tumors, they are frequently dysfunctional in situ. By performing both whole-genome CRISPR and metabolic inhibitor screens, we identify that nicotinamide phosphoribosyltransferase (NAMPT) is required for T cell activation. NAMPT is low in TILs, and its expression is controlled by the transcriptional factor Tubby (TUB), whose activity depends on the T cell receptor-phospholipase C gamma (TCR-PLCγ) signaling axis. The intracellular level of NAD+, whose synthesis is dependent on the NAMPT-mediated salvage pathway, is also decreased in TILs. Liquid chromatography-mass spectrometry (LC-MS) and isotopic labeling studies confirm that NAD+ depletion led to suppressed glycolysis, disrupted mitochondrial function, and dampened ATP synthesis. Excitingly, both adoptive CAR-T and anti-PD1 immune checkpoint blockade mouse models demonstrate that NAD+ supplementation enhanced the tumor-killing efficacy of T cells. Collectively, this study reveals that an impaired TCR-TUB-NAMPT-NAD+ axis leads to T cell dysfunction in the tumor microenvironment, and an over-the-counter nutrient supplement of NAD+ could boost T-cell-based immunotherapy.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse