MilliporeSigma
Cortical gamma-band resonance preferentially transmits coherent input.
Cell reports (2021-05-06)
Christopher Murphy Lewis, Jianguang Ni, Thomas Wunderle, Patrick Jendritza, Andreea Lazar, Ilka Diester, Pascal Fries
ABSTRACT
Synchronization has been implicated in neuronal communication, but causal evidence remains indirect. We use optogenetics to generate depolarizing currents in pyramidal neurons of the cat visual cortex, emulating excitatory synaptic inputs under precise temporal control, while measuring spike output. The cortex transforms constant excitation into strong gamma-band synchronization, revealing the well-known cortical resonance. Increasing excitation with ramps increases the strength and frequency of synchronization. Slow, symmetric excitation profiles reveal hysteresis of power and frequency. White-noise input sequences enable causal analysis of network transmission, establishing that the cortical gamma-band resonance preferentially transmits coherent input components. Models composed of recurrently coupled excitatory and inhibitory units uncover a crucial role of feedback inhibition and suggest that hysteresis can arise through spike-frequency adaptation. The presented approach provides a powerful means to investigate the resonance properties of local circuits and probe how these properties transform input and shape transmission.
Sigma-Aldrich
Anti-GABA Antibody, from rabbit, purified by affinity chromatography