MilliporeSigma
  • Rejuvenated Vintage Tissue Sections Highlight Individual Antigen Fate During Processing and Long-term Storage.
Rejuvenated Vintage Tissue Sections Highlight Individual Antigen Fate During Processing and Long-term Storage.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (2021-09-21)
Francesco Mascadri, Maddalena M Bolognesi, Daniela Pilla, Giorgio Cattoretti
ABSTRACT
Antigen-bearing proteins become progressively unavailable to immunodetection after prolonged storage of routine sections, exposed to a variety of agents, such as moisture, oxygen, and temperature. By proteomic analysis, the antigens are retained in the sections and definitely in the tissue block, pointing to fixation-independent, storage time-dependent protein modifications. Based on previous experience, we hypothesized that a combined exposure to a reducing agent and to chemicals favoring protein conformation changes would reverse the masking in aged sections. Disaccharides, lactose and sucrose, and a surfactant, added to a standard antigen retrieval buffer, reverse the negative changes in aged sections. Furthermore, they provide enhanced access to antigens in freshly cut sections, but not universally, revealing additional factors, besides heat and calcium chelation, required for antigen retrieval of individual proteins.
Sigma-Aldrich
Monoclonal Anti-MKI67 antibody produced in mouse, clone 4A1, ascites fluid
Sigma-Aldrich
Anti-CD14 antibody produced in rabbit, Ab3, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-SOX9 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0639, purified immunoglobulin, buffered aqueous glycerol solution