MilliporeSigma
  • The deubiquitinase OTUD3 stabilizes ACTN4 to drive growth and metastasis of hepatocellular carcinoma.
The deubiquitinase OTUD3 stabilizes ACTN4 to drive growth and metastasis of hepatocellular carcinoma.
Aging (2021-08-13)
Peiyi Xie, Yanglin Chen, Hongfei Zhang, Guichao Zhou, Qing Chao, Jiangwen Wang, Yue Liu, Jiayu Fang, Jing Xie, Jing Zhen, Zhiyuan Wang, Liang Hao, Da Huang
ABSTRACT
OTU domain-containing protein 3 (OTUD3), a deubiquitinating enzyme, has been shown to participate in progression of multiple malignancies. The accurate function of OTUD3 in hepatocellular carcinoma (HCC) progression remains elusive. We found that OTUD3 was significantly overexpressed in HCC, and higher OTUD3 expression was correlated with larger tumor size, more distant metastasis, and worse TNM stage. A series of gain- and loss-of-function assays were also performed to examine the oncogenic function of OTUD3 in promoting HCC cell growth and metastasis in vitro. Using a xenograft mouse model, we showed that OTUD3 accelerated HCC progression in vivo. Furthermore, alpha-actinin 4 (ACTN4) was identified as a downstream target of OTUD3 through mass spectrometry analysis, and the ACTN4 protein level was significantly related to OTUD3 expression. Additionally, OTUD3 directly bound with ACTN4 and deubiquitinated ACTN4 to stabilize it. Finally, ACTN4 was found to be essential for OTUD3-mediated HCC proliferation and metastasis in vitro and in vivo. Collectively, our findings identify the oncogenic role of OTUD3 in HCC and suggest that OTUD3 can be considered as a pivotal prognostic biomarker and a potential therapeutic target.