MilliporeSigma
miR-21 mimic blocks obesity in mice: A novel therapeutic option.
Molecular therapy. Nucleic acids (2021-09-24)
Said Lhamyani, Adriana-Mariel Gentile, Rosa M Giráldez-Pérez, Mónica Feijóo-Cuaresma, Silvana Yanina Romero-Zerbo, Mercedes Clemente-Postigo, Hatem Zayed, Wilfredo Oliva Olivera, Francisco Javier Bermúdez-Silva, Julián Salas, Carlos López Gómez, Abdelkrim Hmadcha, Nabil Hajji, Gabriel Olveira, Francisco J Tinahones, Rajaa El Bekay
ABSTRACT
MicroRNAs (miRNAs) are promising drug targets for obesity and metabolic disorders. Recently, miRNA mimics are providing a unique mechanism of action that guides the process for drug development and sets out the context of their therapeutic application. miRNA (miR)-21 expression in white adipose tissue (WAT) has been associated with obesity. We aimed to analyze miR-21 expression levels in relation to diabetes and obesity to determine the effect that miR-21 mimic has on processes involved in WAT functionality, to dissect the underlying molecular mechanisms, and to study the potential therapeutic application of the miR-21 mimic against obesity. We found higher miR-21 levels in WAT from non-diabetic obese compared to normoweight humans and mice. Moreover, in 3T3-L1 adipocytes, miR-21 mimic affect genes involved in WAT functionality regulation and significantly increase the expression of genes involved in browning and thermogenesis. Interestingly, in vivo treatment with the miR-21 mimic blocked weight gain induced by a high-fat diet in obese mice, without modifying food intake or physical activity. This was associated with metabolic enhancement, WAT browning, and brown adipose tissue (AT) thermogenic programming through vascular endothelial growth factor A (VEGF-A), p53, and transforming growth factor β1 (TGF-β1) signaling pathways. Our findings suggest that miR-21 mimic-based therapy may provide a new opportunity to therapeutically manage obesity and consequently, its associated alterations.
Sigma-Aldrich
Anti-UCP1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Toluidine Blue, 8.74% (ZN (THEORY)), for microscopy (Hist., Vit.)
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)−FITC antibody produced in sheep, affinity isolated antibody, buffered aqueous solution