MilliporeSigma
  • Nicotinamide Riboside Enhances Endothelial Precursor Cell Function to Promote Refractory Wound Healing Through Mediating the Sirt1/AMPK Pathway.
Nicotinamide Riboside Enhances Endothelial Precursor Cell Function to Promote Refractory Wound Healing Through Mediating the Sirt1/AMPK Pathway.
Frontiers in pharmacology (2021-06-01)
Zhen-Hua Wang, Xiao-Gang Bao, Jun-Jie Hu, Si-Bo Shen, Guo-Hua Xu, Ye-Lin Wu
ABSTRACT
Lack of vascularization is directly associated with refractory wound healing in diabetes mellitus (DM). Enrichment of endothelial precursor cells (EPCs) is a promising but challenging approach for the treatment of diabetic wounds. Herein, we investigate the action of nicotinamide riboside (NR) on EPC function for improved healing of diabetic wounds. Db/db mice that were treated with NR-supplemented food (400 mg/kg/d) for 12 weeks exhibited higher wound healing rates and angiogenesis than untreated db/db mice. In agreement with this phenotype, NR supplementation significantly increased the number of blood EPCs and bone marrow (BM)-derived EPCs of db/db mice, as well as the tube formation and adhesion functions of BM-EPCs. Furthermore, NR-supplemented BM-EPCs showed higher expression of sirtuin 1 (Sirt1), phosphorylated adenosine monophosphate-activated protein kinase (p-AMPK), and lower expression of acetylated peroxisome proliferator-activated receptor γ coactivator (PGC-1α) than BM-EPCs isolated from untreated db/db mice. Knockdown of Sirt1 in BM-EPCs significantly abolished the tube formation and adhesion function of NR as well as the expression of p-AMPK and deacetylated PGC-1a. Inhibition of AMPK abolished the NR-regulated EPC function but had no effect on Sirt1 expression, demonstrating that NR enhances EPC function through the Sirt1-AMPK pathway. Overall, this study demonstrates that the oral uptake of NR enhances the EPC function to promote diabetic wound healing, indicating that NR supplementation might be a promising strategy to prevent the progression of diabetic complications.
Sigma-Aldrich
Monoclonal Anti-GAPDH−Peroxidase antibody produced in mouse, clone GAPDH-71.1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-PGC-1 alpha, from rabbit, purified by affinity chromatography