MilliporeSigma
  • Sox6 expression distinguishes dorsally and ventrally biased dopamine neurons in the substantia nigra with distinctive properties and embryonic origins.
Sox6 expression distinguishes dorsally and ventrally biased dopamine neurons in the substantia nigra with distinctive properties and embryonic origins.
Cell reports (2021-11-11)
Milagros Pereira Luppi, Maite Azcorra, Giuliana Caronia-Brown, Jean-Francois Poulin, Zachary Gaertner, Serafin Gatica, Oscar Andrés Moreno-Ramos, Navid Nouri, Marilyn Dubois, Yongchao C Ma, Charu Ramakrishnan, Lief Fenno, Yoon Seok Kim, Karl Deisseroth, Francesca Cicchetti, Daniel A Dombeck, Rajeshwar Awatramani
ABSTRACT
Dopamine (DA) neurons in the ventral tier of the substantia nigra pars compacta (SNc) degenerate prominently in Parkinson's disease, while those in the dorsal tier are relatively spared. Defining the molecular, functional, and developmental characteristics of each SNc tier is crucial to understand their distinct susceptibility. We demonstrate that Sox6 expression distinguishes ventrally and dorsally biased DA neuron populations in the SNc. The Sox6+ population in the ventral SNc includes an Aldh1a1+ subset and is enriched in gene pathways that underpin vulnerability. Sox6+ neurons project to the dorsal striatum and show activity correlated with acceleration. Sox6- neurons project to the medial, ventral, and caudal striatum and respond to rewards. Moreover, we show that this adult division is encoded early in development. Overall, our work demonstrates a dual origin of the SNc that results in DA neuron cohorts with distinct molecular profiles, projections, and functions.
Sigma-Aldrich
Monoclonal Anti-Calbindin-D-28K antibody produced in mouse, clone CB-955, ascites fluid
Sigma-Aldrich
Anti-LMX-1 Antibody, serum, from rabbit
Sigma-Aldrich
Monoclonal Anti-ALDH1A1 antibody produced in mouse, clone 5A11, ascites fluid
Sigma-Aldrich
Anti-SOX6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Triton X-100, for molecular biology
Sigma-Aldrich
Donkey serum
Sigma-Aldrich
Tamoxifen, ≥99%