MilliporeSigma
  • Dimethylglycine and chemically related amines tested for mutagenicity under potential nitrosation conditions.
Dimethylglycine and chemically related amines tested for mutagenicity under potential nitrosation conditions.
Mutation research (1989-04-01)
A J Hoorn
ABSTRACT
Dimethylglycine (DMG) and the chemically related amino acids glycine, sarcosine (monomethylglycine) and betaine (trimethylglycine) were tested in Salmonella typhimurium strain TA100 after treatment with sodium nitrite under acidic conditions using a modified Ames Salmonella/microsome assay as reported by Colman et al. (1980). The increase in the number of revertants observed both with and without metabolic activation was also induced in the control mixtures without adding the amines. From the subsequent testing of the individual components of the mixtures, we concluded that non-consumed nitrite was responsible for the mutagenic responses observed in the different reaction mixtures, and not the amines themselves. There were no consistent indications of mutagenic activity of the DMG test mixture as compared to the control mixture which exhibited both consistent mutagenic activity and a toxic effect which was not increased by the addition of DMG. In fact, DMG seemed to decrease the toxicity of the control reaction solution to the Salmonella which was clearly observed at the higher doses. DMG cannot be considered mutagenic under the test conditions employed. The same can be said of the other amino acids as well.
Sigma-Aldrich
N,N-Dimethylglycine, ≥99%
Sigma-Aldrich
N,N-Dimethylglycine hydrochloride