MilliporeSigma
  • Circadian neurons in the paraventricular nucleus entrain and sustain daily rhythms in glucocorticoids.
Circadian neurons in the paraventricular nucleus entrain and sustain daily rhythms in glucocorticoids.
Nature communications (2021-10-03)
Jeff R Jones, Sneha Chaturvedi, Daniel Granados-Fuentes, Erik D Herzog
ABSTRACT
Signals from the central circadian pacemaker, the suprachiasmatic nucleus (SCN), must be decoded to generate daily rhythms in hormone release. Here, we hypothesized that the SCN entrains rhythms in the paraventricular nucleus (PVN) to time the daily release of corticosterone. In vivo recording revealed a critical circuit from SCN vasoactive intestinal peptide (SCNVIP)-producing neurons to PVN corticotropin-releasing hormone (PVNCRH)-producing neurons. PVNCRH neurons peak in clock gene expression around midday and in calcium activity about three hours later. Loss of the clock gene Bmal1 in CRH neurons results in arrhythmic PVNCRH calcium activity and dramatically reduces the amplitude and precision of daily corticosterone release. SCNVIP activation reduces (and inactivation increases) corticosterone release and PVNCRH calcium activity, and daily SCNVIP activation entrains PVN clock gene rhythms by inhibiting PVNCRH neurons. We conclude that daily corticosterone release depends on coordinated clock gene and neuronal activity rhythms in both SCNVIP and PVNCRH neurons.
Sigma-Aldrich
Anti-BMAL1 Antibody, serum, from guinea pig