MilliporeSigma
  • Soluble Receptor for Advanced Glycation End-products regulates age-associated Cardiac Fibrosis.
Soluble Receptor for Advanced Glycation End-products regulates age-associated Cardiac Fibrosis.
International journal of biological sciences (2021-07-31)
Francesco Scavello, Filippo Zeni, Giuseppina Milano, Federica Macrì, Stefania Castiglione, Estella Zuccolo, Alessandro Scopece, Giovanni Pezone, Calogero C Tedesco, Patrizia Nigro, Genny Degani, Elisa Gambini, Fabrizio Veglia, Laura Popolo, Giulio Pompilio, Gualtiero I Colombo, Marco E Bianchi, Angela Raucci
ABSTRACT
Myocardial aging increases the cardiovascular risk in the elderly. The Receptor for Advanced Glycation End-products (RAGE) is involved in age-related disorders. The soluble isoform (sRAGE) acts as a scavenger blocking the membrane-bound receptor activation. This study aims at investigating RAGE contribution to age-related cardiac remodeling. We analyzed the cardiac function of three different age groups of female Rage-/- and C57BL/6N (WT) mice: 2.5- (Young), 12- (Middle-age, MA) and 21-months (Old) old. While aging, Rage-/- mice displayed an increase in left ventricle (LV) dimensions compared to age-matched WT animals, with the main differences observed in the MA groups. Rage-/- mice showed higher fibrosis and a larger number of α-Smooth Muscle Actin (SMA)+ cells with age, along with increased expression of pro-fibrotic Transforming Growth Factor (TGF)-β1 pathway components. RAGE isoforms were undetectable in LV of WT mice, nevertheless, circulating sRAGE declined with aging and inversely associated with LV diastolic dimensions. Human cardiac fibroblasts stimulated with sRAGE exhibited a reduction in proliferation, pro-fibrotic proteins and TGF-beta Receptor 1 (TGFbR1) expression and Smad2-3 activation. Finally, sRAGE administration to MA WT animals reduced cardiac fibrosis. Hence, our work shows that RAGE associates with age-dependent myocardial changes and indicates sRAGE as an inhibitor of cardiac fibroblasts differentiation and age-dependent cardiac fibrosis.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use in purification of Histidine-tagged proteins, DMSO solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-TGF-beta Receptor Antibody, type I, from rabbit, purified by affinity chromatography