MilliporeSigma
  • Comparison of immunohistochemical staining of a mitochondrial protein, lipoamide dehydrogenase, with Fab'-peroxidase conjugates prepared by maleimide or periodate.
Comparison of immunohistochemical staining of a mitochondrial protein, lipoamide dehydrogenase, with Fab'-peroxidase conjugates prepared by maleimide or periodate.
In vivo (Athens, Greece) (1987-07-01)
S Matuda, Y Yagi, T Saheki, S Hashida, E Ishikawa
ABSTRACT
IgG-maleimide peroxidase, Fab'-maleimide peroxidase, polymer and monomer types of Fab'-periodate peroxidase were prepared from an antibody against rat lipoamide dehydrogenase, a component of the pyruvate dehydrogenase complex which is located in mitochondria. They were examined for immunohistochemical staining of the rat kidney. Fab'-maleimide peroxidase was the best for staining mitochondrial protein. IgG-maleimide peroxidase and the monomer type of Fab'-periodate peroxidase had the same intensity of staining. The polymer type of Fab'-periodate peroxidase could not stain the lipoamide dehydrogenase.
Sigma-Aldrich
Peroxidase, Maleimide Activated from horseradish, lyophilized powder, >200 units/mg protein