MilliporeSigma
  • Role of glutamate dehydrogenase in ammonia assimilation in nitrogen-fixing Bacillus macerans.
Role of glutamate dehydrogenase in ammonia assimilation in nitrogen-fixing Bacillus macerans.
Journal of bacteriology (1987-10-01)
K Kanamori, R L Weiss, J D Roberts
ABSTRACT
Pathways of ammonia assimilation into glutamic acid in Bacillus macerans were investigated by measurements of the specific activities of glutamate dehydrogenase (GDH), glutamine synthetase, and glutamate synthase. In ammonia-rich medium, GDH was the predominant pathway of ammonia assimilation. In nitrogen-fixing cells in which the intracellular NH4+ concentration was 1.4 +/- 0.5 mM, the activity of GDH with a Km of 2.2 mM for NH4+ was found to be severalfold higher than that of glutamate synthase. The result suggests that GDH plays a significant role in the assimilation of NH4+ in N2-fixing B. macerans.
Sigma-Aldrich
NADH Oxidase from Bacillus licheniformis, lyophilized powder