MilliporeSigma
  • HES and Mox genes are expressed during early mesoderm formation in a mollusk with putative ancestral features.
HES and Mox genes are expressed during early mesoderm formation in a mollusk with putative ancestral features.
Scientific reports (2021-09-11)
Attila Sachslehner, Elisabeth Zieger, Andrew Calcino, Andreas Wanninger
ABSTRACT
The mesoderm is considered the youngest of the three germ layers. Although its morphogenesis has been studied in some metazoans, the molecular components underlying this process remain obscure for numerous phyla including the highly diverse Mollusca. Here, expression of Hairy and enhancer of split (HES), Mox, and myosin heavy chain (MHC) was investigated in Acanthochitona fascicularis, a representative of Polyplacophora with putative ancestral molluscan features. While AfaMHC is expressed throughout myogenesis, AfaMox1 is only expressed during early stages of mesodermal band formation and in the ventrolateral muscle, an autapomorphy of the polyplacophoran trochophore. Comparing our findings to previously published data across Metazoa reveals Mox expression in the mesoderm in numerous bilaterians including gastropods, polychaetes, and brachiopods. It is also involved in myogenesis in molluscs, annelids, tunicates, and craniates, suggesting a dual role of Mox in mesoderm and muscle formation in the last common bilaterian ancestor. AfaHESC2 is expressed in the ectoderm of the polyplacophoran gastrula and later in the mesodermal bands and in putative neural tissue, whereas AfaHESC7 is expressed in the trochoblasts of the gastrula and during foregut formation. This confirms the high developmental variability of HES gene expression and demonstrates that Mox and HES genes are pleiotropic.
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Roche
First Strand cDNA Synthesis Kit for RT-PCR (AMV), sufficient for 30 reactions (including 5 control reactions), kit of 1 (10 components), suitable for RT-PCR, hotstart: no, dNTPs included
Roche
BCIP, solution, >95% (HPLC), pkg of 3 mL (150 mg)
Roche
DIG RNA Labeling Mix, sufficient for 20 reactions, solution
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)
Sigma-Aldrich
Dextran sulfate sodium salt from Leuconostoc spp., for molecular biology, average Mw >500,000 (dextran starting material), contains 0.5-2% phosphate buffer
Sigma-Aldrich
Maleic acid, ReagentPlus®, ≥99% (HPLC)
Roche
NBT, 4-Nitro blue tetrazolium chloride, solution
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%