MilliporeSigma
  • SCASP: A Simple and Robust SDS-Aided Sample Preparation Method for Proteomic Research.
SCASP: A Simple and Robust SDS-Aided Sample Preparation Method for Proteomic Research.
Molecular & cellular proteomics : MCP (2021-02-08)
Guohong Gan, Xiao Xu, Xi Chen, Xiu-Fang Zhang, Jinling Wang, Chuan-Qi Zhong
ABSTRACT
SDS is widely used in sample preparation for proteomic research. However, SDS is incompatible with LC and electrospray ionization. SDS depletion is therefore required ahead of LC-MS analysis. Most of current SDS removal strategies are time consuming, laborious, and have low reproducibility. Here, we describe a method, SDS-cyclodextrin (CD)-assisted sample preparation, by which CD can bind to SDS and form CD-SDS complexes in solutions, allowing for direct tryptic digestion. We demonstrate that SDS-CD-assisted sample preparation is a simple, fast, and robust SDS-based sample preparation method for proteomics application.
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
Sigma-Aldrich
Trypsin from porcine pancreas, Proteomics Grade, BioReagent, Dimethylated
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%